ENA238712113 Enamine

5-(methylsulfamoyl)-2-(thiophen-2-yl)benzoic acid

  • MDL number MFCD09042453
  • Cas Number : 926254-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44241

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44241
Availability details for
Quantity
  • Availability
  • Add to Cart