ENA238712116 Enamine

3',4'-dichloro-4-(methylsulfamoyl)-[1,1'-biphenyl]-2-carboxylic acid

  • MDL number MFCD09042489
  • Cas Number : 926225-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44243

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44243
Availability details for
Quantity
  • Availability
  • Add to Cart