ENA238712123 Enamine

3,4-dihydro-2H-pyran-2-carbaldehyde

  • MDL number MFCD00059170
  • Cas Number : 100-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50137

Safety Information

signalword Warning
safety_statement 16-26-33-36/37/39-45
msds https://www.enaminestore.com/catalog/EN300-50137
pictograms GHS02,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart