ENA238712135 Enamine

2-[2-(5-bromothiophen-2-yl)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD11179462
  • Cas Number : 1094355-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50165

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50165
Availability details for
Quantity
  • Availability
  • Add to Cart