ENA238712139 Enamine

2-[(1,3-benzothiazol-2-yl)(methyl)amino]acetic acid

  • MDL number MFCD09729971
  • Cas Number : 104344-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44249
Availability details for
Quantity
  • Availability
  • Add to Cart