ENA238712273 Enamine

1-benzyl-6-oxo-2-phenylpiperidine-3-carboxylic acid

  • MDL number MFCD07377824
  • Cas Number : 94655-32-0
  • Molecular Weight : 309.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50212

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50212
Availability details for
Quantity
  • Availability
  • Add to Cart