ENA238712277 Enamine

ethyl 2-(1-methyl-1H-imidazol-2-yl)acetate hydrochloride

  • MDL number MFCD12912821
  • Cas Number : 1193388-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50259

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50259
Availability details for
Quantity
  • Availability
  • Add to Cart