ENA238712299 Enamine

5-methyl-2,3,4,5-tetrahydro-1,4-benzoxazepine hydrochloride

  • MDL number MFCD12912858
  • Cas Number : 1193389-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164...166
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50498

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50498
Availability details for
Quantity
  • Availability
  • Add to Cart