ENA238712307 Enamine

methyl 5-sulfamoylthiophene-2-carboxylate

  • MDL number MFCD12912868
  • Cas Number : 59777-76-3
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50535

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50535
Availability details for
Quantity
  • Availability
  • Add to Cart