ENA238712310 Enamine

7-(chlorosulfonyl)-2,3-dihydro-1H-indene-5-carboxylic acid

  • MDL number MFCD12912871
  • Cas Number : 1183437-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-50582

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50582
Availability details for
Quantity
  • Availability
  • Add to Cart