ENA238712316 Enamine

2-phenyl-5,6,7,8-tetrahydroquinazolin-6-amine dihydrochloride

  • MDL number MFCD12912881
  • Cas Number : 1193389-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 298...300
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50642

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50642
Availability details for
Quantity
  • Availability
  • Add to Cart