ENA238712318 Enamine

methyl 8-chloro-[1,2,4]triazolo[3,4-a]pyridine-3-carboxylate

  • MDL number MFCD12912882
  • Cas Number : 1193387-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52...55
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50662

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50662
Availability details for
Quantity
  • Availability
  • Add to Cart