ENA238712319 Enamine

2-chloro-6-methoxyquinoline-5-sulfonyl chloride

  • MDL number MFCD12912883
  • Cas Number : 1193387-35-7
  • Molecular Weight : 292.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-50663

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50663
Availability details for
Quantity
  • Availability
  • Add to Cart