ENA238712320 Enamine

methyl 2-{[4-(aminomethyl)phenyl]formamido}acetate hydrochloride

  • MDL number MFCD12912885
  • Cas Number : 1193387-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50673
Availability details for
Quantity
  • Availability
  • Add to Cart