ENA238712321 Enamine

3-cyclohexyl-1-(piperazin-1-yl)propan-1-one hydrochloride

  • MDL number MFCD12912887
  • Cas Number : 1193390-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50892

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50892
Availability details for
Quantity
  • Availability
  • Add to Cart