ENA238712323 Enamine

5-(piperazin-1-ylcarbonyl)-1H-1,3-benzodiazole dihydrochloride

  • MDL number MFCD12912889
  • Cas Number : 1193390-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 314...316
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50921

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50921
Availability details for
Quantity
  • Availability
  • Add to Cart