ENA238712332 Enamine

N,2,3-trimethylaniline hydrochloride

  • MDL number MFCD12912898
  • Cas Number : 1193388-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50968

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50968
Availability details for
Quantity
  • Availability
  • Add to Cart