ENA238712347 Enamine

2-(2,5-dimethylpyrrolidin-1-yl)acetic acid

  • MDL number MFCD12912924
  • Cas Number : 1193388-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51113

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51113
Availability details for
Quantity
  • Availability
  • Add to Cart