ENA238712349 Enamine

8-benzyl-8-azabicyclo[4.3.1]decan-10-one

  • MDL number MFCD13544605
  • Cas Number : 936110-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51135

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51135
Availability details for
Quantity
  • Availability
  • Add to Cart