ENA238712366 Enamine

1-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbothioamide

  • MDL number MFCD12912967
  • Cas Number : 1193388-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-51454

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51454
Availability details for
Quantity
  • Availability
  • Add to Cart