ENA238712368 Enamine

tert-butyl 4-(pyrrolidine-2-carbonyl)piperazine-1-carboxylate

  • MDL number MFCD12912976
  • Cas Number : 1219201-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-51513

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51513
Availability details for
Quantity
  • Availability
  • Add to Cart