ENA238712371 Enamine

N-[4-(chloromethyl)-1,3-thiazol-2-yl]-N-(4-fluorophenyl)acetamide

  • MDL number MFCD12912980
  • Cas Number : 1193390-14-5
  • Molecular Weight : 284.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51533

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51533
Availability details for
Quantity
  • Availability
  • Add to Cart