ENA238712372 Enamine

N-cyclopentyl-N,3,5-trimethyl-1H-pyrazole-4-sulfonamide hydrochloride

  • MDL number MFCD12912982
  • Cas Number : 1231928-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51546

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51546
Availability details for
Quantity
  • Availability
  • Add to Cart