ENA238712375 Enamine

2,6-difluoro-N-methylaniline hydrochloride

  • MDL number MFCD12912996
  • Cas Number : 1193390-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51661

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51661
Availability details for
Quantity
  • Availability
  • Add to Cart