ENA238712381 Enamine

5-(propan-2-yloxy)-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD12913005
  • Cas Number : 37158-92-2
  • Molecular Weight : 176.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51712

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51712
Availability details for
Quantity
  • Availability
  • Add to Cart