ENA238712382 Enamine

(2S,4R)-N-[(1R,2R)-2-hydroxy-1-[(2R,3R,5R,6R)-3,4,5-trihydroxy-6-(methylsulfanyl)oxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide hydrochloride

  • MDL number MFCD12913006
  • Cas Number : 859-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51718

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51718
s_phrases_eu 22-24/25
Availability details for
Quantity
  • Availability
  • Add to Cart