ENA238712387 Enamine

4-fluoro-3-methanesulfonylbenzene-1-sulfonyl chloride

  • MDL number MFCD12913017
  • Cas Number : 1188525-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51814

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51814
Availability details for
Quantity
  • Availability
  • Add to Cart