ENA238712390 Enamine

tert-butyl N-(carbamimidoylmethyl)carbamate hydrochloride

  • MDL number MFCD12913031
  • Cas Number : 691898-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188...190
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51899

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-51899
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart