ENA238712393 Enamine

3-(4-bromothiophen-2-yl)-5-(chloromethyl)-1,2,4-oxadiazole

  • MDL number MFCD12913040
  • Cas Number : 1193388-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-51973

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51973
Availability details for
Quantity
  • Availability
  • Add to Cart