ENA238712398 Enamine

2-{2-[(cyclopropylcarbamoyl)amino]-1,3-thiazol-4-yl}acetic acid

  • MDL number MFCD12913047
  • Cas Number : 1193387-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52022

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52022
Availability details for
Quantity
  • Availability
  • Add to Cart