ENA238712401 Enamine

2-amino-N-(4-hydroxycyclohexyl)acetamide hydrochloride

  • MDL number MFCD12913051
  • Cas Number : 1193389-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52035

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52035
Availability details for
Quantity
  • Availability
  • Add to Cart