ENA238712417 Enamine

1-(1,3-dimethyl-1H-pyrazol-4-yl)ethan-1-amine

  • MDL number MFCD03422567
  • Cas Number : 911788-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50198

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50198
Availability details for
Quantity
  • Availability
  • Add to Cart