ENA238712422 Enamine

1-phenyl-3-(propan-2-yl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD11180136
  • Cas Number : 874908-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59...61
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50463

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50463
Availability details for
Quantity
  • Availability
  • Add to Cart