ENA238712424 Enamine

2-phenyl-5,6,7,8-tetrahydroquinazolin-6-amine

  • MDL number MFCD08447512
  • Cas Number : 929973-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50465

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50465
Availability details for
Quantity
  • Availability
  • Add to Cart