ENA238712427 Enamine

2-(chloromethyl)-6-fluoro-1H-1,3-benzodiazole-4-carboxylic acid hydrochloride

  • MDL number MFCD12912813
  • Cas Number : 1193389-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50202

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50202
Availability details for
Quantity
  • Availability
  • Add to Cart