ENA238712429 Enamine

5-(chlorosulfonyl)benzene-1,3-dicarboxylic acid

  • MDL number MFCD00090839
  • Cas Number : 134178-04-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-50470

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50470
Availability details for
Quantity
  • Availability
  • Add to Cart