ENA238712438 Enamine

4-ethyl-5-methyl-1,3-thiazol-2-amine

  • MDL number MFCD07186525
  • Cas Number : 30709-59-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50486

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50486
Availability details for
Quantity
  • Availability
  • Add to Cart