ENA238712439 Enamine

4-(1-aminoethyl)-N,N-dimethylbenzene-1-sulfonamide hydrochloride

  • MDL number MFCD12912814
  • Cas Number : 1193389-49-9
  • Molecular Weight : 264.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-50225

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50225
Availability details for
Quantity
  • Availability
  • Add to Cart