ENA238712454 Enamine

1-benzyl-1H-pyrazol-4-amine

  • MDL number MFCD01693690
  • Cas Number : 28466-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50177
Availability details for
Quantity
  • Availability
  • Add to Cart