ENA238712459 Enamine

1-chloro-3-(2-chloro-6-fluorophenyl)propan-2-one

  • MDL number MFCD11621969
  • Cas Number : 1094902-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 37...39
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50512

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50512
Availability details for
Quantity
  • Availability
  • Add to Cart