ENA238712462 Enamine

(prop-2-en-1-yl)(propan-2-yl)amine hydrochloride

  • MDL number MFCD12912817
  • Cas Number : 50529-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-50233

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50233
Availability details for
Quantity
  • Availability
  • Add to Cart