ENA238712464 Enamine

N-(4-amino-2-methylphenyl)-2-(1H-pyrazol-1-yl)acetamide dihydrochloride

  • MDL number MFCD12912864
  • Cas Number : 1193389-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50521

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50521
Availability details for
Quantity
  • Availability
  • Add to Cart