ENA238712467 Enamine

4-(5-amino-4-cyano-1H-pyrazol-1-yl)benzoic acid

  • MDL number MFCD04610186
  • Cas Number : 882233-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50528

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50528
Availability details for
Quantity
  • Availability
  • Add to Cart