ENA238712469 Enamine

1-(piperidin-4-yl)-1H,4H,5H,6H,7H-[1,2,3]triazolo[4,5-d]pyridazine-4,7-dione hydrochloride

  • MDL number MFCD12912818
  • Cas Number : 1193388-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-50235

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50235
Availability details for
Quantity
  • Availability
  • Add to Cart