ENA238712474 Enamine

3-(morpholin-4-yl)aniline

  • MDL number MFCD03197165
  • Cas Number : 159724-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113...115
Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50538

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50538
Availability details for
Quantity
  • Availability
  • Add to Cart