ENA238712476 Enamine

3-(dimethyl-1,3-oxazol-2-yl)aniline

  • MDL number MFCD06739710
  • Cas Number : 827582-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-50540

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50540
Availability details for
Quantity
  • Availability
  • Add to Cart