ENA238712479 Enamine

N-(4-amino-2-methylphenyl)ethane-1-sulfonamide

  • MDL number MFCD09042751
  • Cas Number : 926246-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50544
Availability details for
Quantity
  • Availability
  • Add to Cart