ENA238712480 Enamine

5-fluoro-2-(3-methoxypropanamido)benzoic acid

  • MDL number MFCD11649320
  • Cas Number : 1099091-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50545

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50545
Availability details for
Quantity
  • Availability
  • Add to Cart