ENA238712484 Enamine

[2-(butan-2-yloxy)phenyl]boronic acid

  • MDL number MFCD11183173
  • Cas Number : 1313760-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-50549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50549
Availability details for
Quantity
  • Availability
  • Add to Cart