ENA238712487 Enamine

[2-(1H-pyrrol-1-yl)thiophen-3-yl]methanamine

  • MDL number MFCD09041787
  • Cas Number : 63647-04-1
  • Molecular Weight : 178.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50552

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50552
Availability details for
Quantity
  • Availability
  • Add to Cart